Learn More
ILC Dover Sentinel XT Non-Incendive Starter Kit
Catalog No. 19631083
Change view
Click to view available options
Includes: Blower, Battery, Flowmeter
Catalog No.
19-631-083 Includes: Blower, Battery, Flowmeter
1 options
Catalog No. 19-631-083 Supplier ILC Dover Supplier No. S3326
Add to Cart
Add to Cart
Starter kit is equipped with the Sentinel XT Non-Incendive Blower (Blue), LiPo rechargeable battery with latch, flow meter, charging cradle, and Quick loc belt.
  • Comes with the Sentinel XT Non-Incendive Blower (Blue), LiPo Rechargeable battery with latch, flowmeter, charging cradle and the Quick loc belt
  • Powered air-purifying respirator assemblies and systems include parts required to build a full system or to replace existing parts
  • Belt-mounted blower distributes ambient air through a filter to provide clean, breathable air to the wearer
  • PAPRs allow workers to move freely and unrestricted
  • System parts are compatible with components of the same brand and series
Specifications
Product Type PAPR Assembly, No Headtop
Includes Sentinel XT Non-Incendive Blower (Blue), LiPo Rechargeable battery with latch, flowmeter, charging cradle and the Quick loc belt
Model Sentinel XT