Learn More
ILC Dover Sentinel XT Starter Kit
Catalog No. 19631084
Change view
Click to view available options
Includes: Blower, Battery, Flowmeter
Catalog No.
19-631-084 Includes: Blower, Battery, Flowmeter
1 options
Catalog No. 19-631-084 Supplier ILC Dover Supplier No. S3327
Add to Cart
Add to Cart
Starter kit is equipped with the Sentinel XT Blower (Gray), LiPo rechargeable battery, flow meter, charging cradle, and Quick loc belt.
  • Comes with the Sentinel XT Blower (Gray), LiPo Rechargeable battery, flowmeter, charging cradle and the Quick loc belt
  • Powered air-purifying respirator assemblies and systems include parts required to build a full system or to replace existing parts
  • Belt-mounted blower distributes ambient air through a filter to provide clean, breathable air to the wearer
  • PAPRs allow workers to move freely and unrestricted
  • System parts are compatible with components of the same brand and series
Specifications
Product Type PAPR Assembly, No Headtop
Includes Sentinel XT Blower (Gray), LiPo Rechargeable battery, flowmeter, charging cradle and the Quick loc belt
Model Sentinel XT