Learn More
Cytiva (Formerly Pall Lab) Sentino Global Membrane Dispenser (1/pkg)
Supplier: Cytiva 13184
Simple design, reliable dispenseEasy to load Simple loading and unloading of refill pack at any point during use.No jamming Guide plates assist loading and keep refill web in alignment during use to avoid jamming of the instrument.Soft touch keypad Aseptically dispense a disc of membrane at the touch of a button.Designed to hold 200 membrane discs Saves time by eliminating interruptions to reload dispenser.Compact design Small footprint frees valuable bench top space and provides flexibility in arranging workspace for optimal efficiency and workflow.Miniaturized refill pack Sized to minimize storage space resulting in less packaging material waste.Portable Use plugged into main power source or charge battery for use anywhere. Indicator lights let you know when the battery charge is running low.Easy to clean Sanitize with standard laboratory disinfectants. This product is not approved for use in medical, clinical, surgical or other patient applications.