Learn More
YSI 3P LifeScience SENTIX 82 PH COMB ELECTRODE
Catalog No. NC9704449
Encompass_Preferred
Glass pH combination electrode with liquid electrolyte, built-in temperature sensor, BNC plug and banana plug, length 120 mm, 3.3 ft (1 m) cable.
Catalog No. NC9704449 Supplier YSI 3P LifeScience Supplier No. 103643
Glass pH combination electrode with liquid electrolyte, built-in temperature sensor, BNC plug and banana plug, length 120 mm, 3.3 ft (1 m) cable.