Learn More
Cayman Chemical SenTraGorCel Senscence Re 10mg
SDP
Catalog No. 502909979
Encompass_Preferred
A PKM inhibitor (Ki 0 076-2 11 UM) inhibits PKCa at 10 UM prevents ionomycin-induced PKC translocation to the plasma membrane in HEK293 cells expressing AKAP79 at 5 UM inhibits the conditioned place preference response to morphine in rats when administered intracranially into the nucleus accumbens core at doses of 10 and 30 nmol/0 5 Ul per side
Catalog No. 50-290-9979 Supplier Cayman Chemical Supplier No. 3556810 MG
A PKM inhibitor (Ki 0 076-2 11 UM) inhibits PKCa at 10 UM prevents ionomycin-induced PKC translocation to the plasma membrane in HEK293 cells expressing AKAP79 at 5 UM inhibits the conditioned place preference response to morphine in rats when administered intracranially into the nucleus accumbens core at doses of 10 and 30 nmol/0 5 Ul per side