Learn More
SENTRONIC US CORP SENTROPAT FO NIR 1150-2100-256
Catalog No. NC3653425
Encompass
SentroPAT FO NIR 1150-2100 / 256 / 1ch / ALCompact NIR process spectrometer with diode array detector for 1150-2100nm spectral range; 1 Channel systemOffer No.: OUSORER2511935
Catalog No. NC3653425 Supplier SENTRONIC US CORP Supplier No. 2530800600141
Add to Cart
Add to Cart
Request a Quote
SentroPAT FO NIR 1150-2100 / 256 / 1ch / ALCompact NIR process spectrometer with diode array detector for 1150-2100nm spectral range; 1 Channel systemOffer No.: OUSORER2511935