Learn More
SENTRONIC US CORP SENTROPROBE DR LS NIR 200 ATEX
Catalog No. NC3653426
Encompass
SentroProbe DR LS NIR 200 ATEXProbe for diffuse reflectance measurement in the NIR range with 2 internal light source and internal reference, probe length 200mm, ATEX zone 20/21 rated, design according to drawing P07000509003
Catalog No. NC3653426 Supplier SENTRONIC US CORP Supplier No. 2530000901116
Add to Cart
Add to Cart
Request a Quote
SentroProbe DR LS NIR 200 ATEXProbe for diffuse reflectance measurement in the NIR range with 2 internal light source and internal reference, probe length 200mm, ATEX zone 20/21 rated, design according to drawing P07000509003