Learn More
Kimberly-Clark™ Professional Sentry III™ Cap
Catalog No. 173501A
Click to view available options
Ratchet; White
Catalog No.
17-350-1A Ratchet; White
1 options
This item is not returnable. View return policy
Catalog No. 17-350-1A Supplier Kimberly-Clark™ Professional Supplier No. 14409
This item is not returnable. View return policy
Low-riding nape strap prevents cap from slipping
  • Low-profile, silhouette-styled cap
  • Lightweight, molded from high-density polyethylene (HDPE)
  • Accessory slots for attaching additional personal protective equipment
  • Six-point ratchet suspension
  • Fits head sizes 61/2 to 8
Compliance
Meets ANSI Z89.1, Class C, G and E
Specifications
Color White
Material HDPE
Suspension Type 6-Point Ratchet