Learn More
Grainger Sentry Plus Shirt Black Neck 17 in
Catalog No. 502752458
Encompass_Preferred
The Grainger 14F634 is a Westward brand 1/4-inch drive socket set. This 108-piece set includes sizes ranging from 4mm to 14mm as well as 1/4, 5/16, 3/8, and 7/16-inch sockets. The chrome vanadium steel sockets have a matte finish and feature laser etched size markings. The set comes organized in a molded case with fold-out trays for easy storage and portability. This socket set is ideal for automotive and machinery applications that require standard and deep well sockets in fractional and metric sizes.
Catalog No. 50-275-2458 Supplier Grainger Supplier No. 14F634
The Grainger 14F634 is a Westward brand 1/4-inch drive socket set. This 108-piece set includes sizes ranging from 4mm to 14mm as well as 1/4, 5/16, 3/8, and 7/16-inch sockets. The chrome vanadium steel sockets have a matte finish and feature laser etched size markings. The set comes organized in a molded case with fold-out trays for easy storage and portability. This socket set is ideal for automotive and machinery applications that require standard and deep well sockets in fractional and metric sizes.