Learn More
Grainger Sentry Plus Shirt Black Neck 19 in
Catalog No. 502749133
Encompass_Preferred
The Grainger 14F653 is a 14-inch, 6-blade, cast aluminum fan blade with a 53-degree pitch for counter-clockwise rotation. It meets AMCA standards and pairs with various Grainger fan motors and housings for industrial ventilation. Its durable aluminum construction and aerodynamic design ensure efficient airflow, making it suitable for medium to large ventilation needs. This blade is an ideal replacement for many existing Grainger fan setups.
Catalog No. 50-274-9133 Supplier Grainger Supplier No. 14F653
The Grainger 14F653 is a 14-inch, 6-blade, cast aluminum fan blade with a 53-degree pitch for counter-clockwise rotation. It meets AMCA standards and pairs with various Grainger fan motors and housings for industrial ventilation. Its durable aluminum construction and aerodynamic design ensure efficient airflow, making it suitable for medium to large ventilation needs. This blade is an ideal replacement for many existing Grainger fan setups.