Learn More
Grainger Sentry Plus Shirt Blue Neck 17-1/2 in
Catalog No. 502753029
Encompass_Preferred
Grainger's PIR motion sensors are passive infrared detectors for indoor or outdoor security, sensing movement and infrared energy changes. They feature adjustable sensitivity, detection range, time delay, and pulse count, offering reliable detection and immunity to RFI and temperature changes. Standard models cover up to 700 sq ft, while long-range models cover up to 1300 sq ft. Easy to install with low current draw, they come in various models with different ranges, coverage, mounting, and voltage options.
Catalog No. 50-275-3029 Supplier Grainger Supplier No. 14F680
Grainger's PIR motion sensors are passive infrared detectors for indoor or outdoor security, sensing movement and infrared energy changes. They feature adjustable sensitivity, detection range, time delay, and pulse count, offering reliable detection and immunity to RFI and temperature changes. Standard models cover up to 700 sq ft, while long-range models cover up to 1300 sq ft. Easy to install with low current draw, they come in various models with different ranges, coverage, mounting, and voltage options.