Learn More
Grainger Sentry Plus Shirt Blue Neck 18 in
Catalog No. 502753441
Encompass_Preferred
The Grainger 14F685 is a Westward brand 1/4-inch drive socket set. It contains both SAE and metric sockets ranging from 4mm to 15mm and 1/8 inch to 9/16 inch. The set has a total of 38 pieces and comes in a plastic organizer case for storage and portability. The sockets are made from chrome vanadium steel for durability and have a matte finish to prevent corrosion. This socket set is ideal for automotive and light industrial applications.
Catalog No. 50-275-3441 Supplier Grainger Supplier No. 14F685
The Grainger 14F685 is a Westward brand 1/4-inch drive socket set. It contains both SAE and metric sockets ranging from 4mm to 15mm and 1/8 inch to 9/16 inch. The set has a total of 38 pieces and comes in a plastic organizer case for storage and portability. The sockets are made from chrome vanadium steel for durability and have a matte finish to prevent corrosion. This socket set is ideal for automotive and light industrial applications.