Learn More
Grainger Sentry Plus Trouser Womens Dark Navy 24
Catalog No. 502749664
Encompass_Preferred
The Tripp Lite SmartRack 14U Open Frame Rack is a 4-post open frame rack that provides 14U of storage space for mounting networking and IT equipment. It is constructed with a steel frame with adjustable mounting rails that have square mounting holes. The rack has a weight capacity of 200 lbs and has a black powder coat finish. The overall dimensions are 24" wide x 27" deep x 28" high. This rack requires assembly and provides an open structure for efficient airflow and cable management in IT closets and data centers.
Catalog No. 50-274-9664 Supplier Grainger Supplier No. 14U614
The Tripp Lite SmartRack 14U Open Frame Rack is a 4-post open frame rack that provides 14U of storage space for mounting networking and IT equipment. It is constructed with a steel frame with adjustable mounting rails that have square mounting holes. The rack has a weight capacity of 200 lbs and has a black powder coat finish. The overall dimensions are 24" wide x 27" deep x 28" high. This rack requires assembly and provides an open structure for efficient airflow and cable management in IT closets and data centers.