Learn More
Grainger Sentry Plus Trouser Womens Dark Navy 10
Catalog No. 502750470
Encompass_Preferred
The Tripp Lite 14U607 is a 19-inch open frame rack that is 60 inches tall. It has a weight capacity of 1,000 lbs and is constructed from steel with a black powder coat finish. The rack has adjustable mounting rails that allow equipment to be mounted at various depths. It can accommodate EIA-standard 19-inch rack equipment. The open frame design allows efficient cabling and cooling. This rack provides an economical solution for housing rackmount equipment.
Catalog No. 50-275-0470 Supplier Grainger Supplier No. 14U607
The Tripp Lite 14U607 is a 19-inch open frame rack that is 60 inches tall. It has a weight capacity of 1,000 lbs and is constructed from steel with a black powder coat finish. The rack has adjustable mounting rails that allow equipment to be mounted at various depths. It can accommodate EIA-standard 19-inch rack equipment. The open frame design allows efficient cabling and cooling. This rack provides an economical solution for housing rackmount equipment.