Learn More
Grainger Sentry Plus Trouser Womens Dark Navy 16
Catalog No. 502750472
Encompass_Preferred
The Tripp Lite SMART1500LCDT 1500VA 900W UPS offers battery backup and surge protection for electronics during outages. It features 8 outlets: 6 with battery backup and surge protection, and 2 surge-only. The LCD displays UPS status and load. Communication options include tel/DSL, USB, DB9 serial, and SNMP, with an optional Webcard for remote monitoring. Its tower design allows flexible placement.
Catalog No. 50-275-0472 Supplier Grainger Supplier No. 14U610
The Tripp Lite SMART1500LCDT 1500VA 900W UPS offers battery backup and surge protection for electronics during outages. It features 8 outlets: 6 with battery backup and surge protection, and 2 surge-only. The LCD displays UPS status and load. Communication options include tel/DSL, USB, DB9 serial, and SNMP, with an optional Webcard for remote monitoring. Its tower design allows flexible placement.