Learn More
Enterprise Technology Solutions Sentry Safe Fire/Water Safe, w/ Digital Alarm, 18.6"x19.3"x23.8", GMBK
SDP
Catalog No. 5000791768
Encompass_Preferred
With water-resistance and fire-resistance, this digital alarm fire safe provides reliable protection for important documents and valuables. It is UL Classified for one-hour fire endurance, UL verified for 15' drop and ETL verified for one-hour digital media protection. Built-in, loud alarm lets owners know of unauthorized access. Dual touch lock and key provide quick access with added security. To help prevent forced entry, the pry-resistant hinge bar and six large, 1" bolts make you feel confident your most important items are protected.
Catalog No. 50-007-91768 Supplier Enterprise Technology Solutions Supplier No. SENSFW205UPC
With water-resistance and fire-resistance, this digital alarm fire safe provides reliable protection for important documents and valuables. It is UL Classified for one-hour fire endurance, UL verified for 15' drop and ETL verified for one-hour digital media protection. Built-in, loud alarm lets owners know of unauthorized access. Dual touch lock and key provide quick access with added security. To help prevent forced entry, the pry-resistant hinge bar and six large, 1" bolts make you feel confident your most important items are protected.