Learn More
BD Sentry™ Safety Lancet
Catalog No. B369523 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Color:
Green
Catalog No. Color
B369523 Green
1 options
This item is not returnable. View return policy
Catalog No. B369523 Supplier BD Supplier No. 369523
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Allows for quick and safe blood collection from the fingertip
  • Activated by the push of a button
  • Mechanism to prevent reuse and needlestick injuries
  • Easy disposal in sharps containers
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Packaging Quantity 100/Pk.
Needle Gauge 23G
Depth (Metric) Needle 1.8 mm
Color Green
Sterility Sterile