Learn More
Grainger Sentry Shirt Navy Neck 20 in
Catalog No. 502750558
Encompass_Preferred
The Grainger 14J136 is a Westward brand 1/4-inch drive socket set. This 108-piece socket set includes both SAE and metric sockets in shallow, deep, and universal depths, ranging from 1/4 inch drive to 3/4 inch drive. The set comes organized in a sturdy plastic blow mold case for convenient storage and portability. This professional-grade socket set is ideal for automotive and machinery repair.
Catalog No. 50-275-0558 Supplier Grainger Supplier No. 14J136
The Grainger 14J136 is a Westward brand 1/4-inch drive socket set. This 108-piece socket set includes both SAE and metric sockets in shallow, deep, and universal depths, ranging from 1/4 inch drive to 3/4 inch drive. The set comes organized in a sturdy plastic blow mold case for convenient storage and portability. This professional-grade socket set is ideal for automotive and machinery repair.