Learn More
Grainger Sentry Shirt Navy Neck 20 in
Catalog No. 502750240
Encompass_Preferred
The Grainger 14J137 is a 14-inch diameter, 137-tooth gear made of cast iron with a pitch diameter of 14 inches. It has a standard 20 degree pressure angle and features a hub projection on one side. The gear has an approximate weight of 22 lbs and operates on a bore size of 1-3/4 inches. It is manufactured in the USA by Martin Sprocket and Gear. This industrial-grade, large cast iron gear is commonly used for power transmission applications.
Catalog No. 50-275-0240 Supplier Grainger Supplier No. 14J137
The Grainger 14J137 is a 14-inch diameter, 137-tooth gear made of cast iron with a pitch diameter of 14 inches. It has a standard 20 degree pressure angle and features a hub projection on one side. The gear has an approximate weight of 22 lbs and operates on a bore size of 1-3/4 inches. It is manufactured in the USA by Martin Sprocket and Gear. This industrial-grade, large cast iron gear is commonly used for power transmission applications.