Learn More
Cayman Chemical SEP-0372814 10mg
SDP
Catalog No. 502948832
Encompass_Preferred
A PDE10A inhibitor (IC50 = ≤10 nM for the recombinant human enzyme)
Catalog No. 50-294-8832 Supplier Cayman Chemical Supplier No. 3283610 MG
A PDE10A inhibitor (IC50 = ≤10 nM for the recombinant human enzyme)