Learn More
Cayman Chemical SEP-0372814, Purity: ≥98%, CAS: 1516895-53-6, 50 mg
SDP
Catalog No. 502948130
Encompass_Preferred
A PDE10A inhibitor (IC50 = ≤10 nM for the recombinant human enzyme)
Catalog No. 50-294-8130 Supplier Cayman Chemical Supplier No. 3283650 MG
A PDE10A inhibitor (IC50 = ≤10 nM for the recombinant human enzyme)