Learn More
TARGETMOL CHEMICALS INC SEP-363856 HYDROCHLORIDE 5MG
SDP
Catalog No. 502581405
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SEP-363856 hydrochloride (SEP-856 hydrochloride) is an agent of orally active and CNS active psychotropic with a unique non-D2/5-HT2A mechanism of action exerts its antipsychotic-like effects has the potential to treatment of schizophrenia. purity: 99%
Catalog No. 50-258-1405 Supplier TARGETMOL CHEMICALS INC Supplier No. T12899L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SEP-363856 hydrochloride (SEP-856 hydrochloride) is an agent of orally active and CNS active psychotropic with a unique non-D2/5-HT2A mechanism of action exerts its antipsychotic-like effects has the potential to treatment of schizophrenia. purity: 99%