Learn More
Chemglass Life Sciences 125mL Separatory Funnel, Squibb, 14/20 Joint Size
SDP
Catalog No. CG174309
Encompass_Preferred
125mL Separatory Funnel, Squibb, 14/20 Joint Size. Squibb style separatory funnel with a PTFE stopcock. Not supplied with stopper. For stoppers see CG-3000. Stopcock bore on all sizes is 2mm.
Catalog No. CG174309 Supplier Chemglass Life Sciences Supplier No. CG174309
125mL Separatory Funnel, Squibb, 14/20 Joint Size. Squibb style separatory funnel with a PTFE stopcock. Not supplied with stopper. For stoppers see CG-3000. Stopcock bore on all sizes is 2mm.