Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Sep-Pak Solid Phase Extraction, 46 mg Sorbent, 40 μm, 300 Å, 50/pk

Catalog No. 502269315 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These silica-based, hydrophilic, strong anion-exchanger cartridges feature a large pore size (300Å) and a carbonate counter-ion, ideal for extracting anionic analytes in aqueous and non-aqueous solutions. The “Light-style” design with reduced internal diameter optimizes recoveries and reduces solvent consumption by allowing elution of fractions in minimal volume.

  • Silica-based, hydrophilic, strong anion-exchanger.
  • 300Å pore size, carbonate counter-ion.
  • Extracts anionic analytes from aqueous and non-aqueous solutions.
  • “Light-style” design for minimal elution volume and improved recoveries.
  • Reduces solvent consumption.
  • Applications: anionic proteins, acidic pigments, phenolic compounds, peptide pool fractionation, inorganic anions.
  • Mass spectrometry compatible and water wettable.
  • pH range: 2 to 8.
  • Hold up volume: 0.15 mL.

This item is not returnable. View return policy
Catalog No. 50-226-9315 Supplier Waters Corp Supplier No. 186000132DL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These silica-based, hydrophilic, strong anion-exchanger cartridges feature a large pore size (300Å) and a carbonate counter-ion, ideal for extracting anionic analytes in aqueous and non-aqueous solutions. The “Light-style” design with reduced internal diameter optimizes recoveries and reduces solvent consumption by allowing elution of fractions in minimal volume.

  • Silica-based, hydrophilic, strong anion-exchanger.
  • 300Å pore size, carbonate counter-ion.
  • Extracts anionic analytes from aqueous and non-aqueous solutions.
  • “Light-style” design for minimal elution volume and improved recoveries.
  • Reduces solvent consumption.
  • Applications: anionic proteins, acidic pigments, phenolic compounds, peptide pool fractionation, inorganic anions.
  • Mass spectrometry compatible and water wettable.
  • pH range: 2 to 8.
  • Hold up volume: 0.15 mL.

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.