Learn More
Waters Corp Waters Cartridge, C18, Double Luer-Lock, 55–105 μm Particle Size
Catalog No. 502269424 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This cartridge features a silica-based bonded phase with strong hydrophobicity, designed to adsorb even weakly hydrophobic analytes from aqueous solutions. Typical applications include the analysis of drugs and their metabolites in serum, plasma, or urine, desalting peptides, identifying trace organics in environmental water samples, and analyzing organic acids in beverages. Its behavior is similar to reversed-phase HPLC columns.
  • Includes double Luer-Lock adaptors.
  • Silica-based bonded phase with strong hydrophobicity.
  • Adsorbs analytes of even weak hydrophobicity from aqueous solutions.
  • Applications include analysis of drugs and their metabolites in serum, plasma, or urine; desalting peptides; identifying trace organics in environmental water samples; and analyzing organic acids in beverages.
  • Exhibits behavior similar to reversed-phase HPLC columns.
This item is not returnable. View return policy
Catalog No. 50-226-9424 Supplier Waters Corp Supplier No. WAT023501DL
This cartridge features a silica-based bonded phase with strong hydrophobicity, designed to adsorb even weakly hydrophobic analytes from aqueous solutions. Typical applications include the analysis of drugs and their metabolites in serum, plasma, or urine, desalting peptides, identifying trace organics in environmental water samples, and analyzing organic acids in beverages. Its behavior is similar to reversed-phase HPLC columns.
  • Includes double Luer-Lock adaptors.
  • Silica-based bonded phase with strong hydrophobicity.
  • Adsorbs analytes of even weak hydrophobicity from aqueous solutions.
  • Applications include analysis of drugs and their metabolites in serum, plasma, or urine; desalting peptides; identifying trace organics in environmental water samples; and analyzing organic acids in beverages.
  • Exhibits behavior similar to reversed-phase HPLC columns.