Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Sep-Pak Vac Cartridge, C18, 500 mg Sorbent, 6 cc Barrel, 55–105 μm Particle Size

Catalog No. 50179648 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Sep-Pak C18 6 cc Vac Cartridge is a silica-based octadecyl bonded phase product designed for solid phase extraction. It effectively adsorbs analytes from aqueous solutions, including weakly hydrophobic ones. Key applications include extraction of drugs, metabolites, peptides, and organics from water and beverages.

  • Sorbent weight: 500 mg
  • Particle size: 55–105 μm
  • Barrel size: 6 cc
  • Hold-up volume: 1.2 mL
  • pH range: 2 to 8
  • Package of 300 units
  • C18 chemistry, reversed phase mode
  • Endcapped and mass spectrometer compatible
  • Monofunctional silane chemistry for high repeatability, recovery, and selectivity
  • Designed for vacuum manifolds and automated SPE
  • High-quality packings with excellent surface area and pore size
  • Non-contaminating poly-foil pouch packaging

Catalog No. 50-179-648 Supplier Waters Corp Supplier No. 186006325
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Sep-Pak C18 6 cc Vac Cartridge is a silica-based octadecyl bonded phase product designed for solid phase extraction. It effectively adsorbs analytes from aqueous solutions, including weakly hydrophobic ones. Key applications include extraction of drugs, metabolites, peptides, and organics from water and beverages.

  • Sorbent weight: 500 mg
  • Particle size: 55–105 μm
  • Barrel size: 6 cc
  • Hold-up volume: 1.2 mL
  • pH range: 2 to 8
  • Package of 300 units
  • C18 chemistry, reversed phase mode
  • Endcapped and mass spectrometer compatible
  • Monofunctional silane chemistry for high repeatability, recovery, and selectivity
  • Designed for vacuum manifolds and automated SPE
  • High-quality packings with excellent surface area and pore size
  • Non-contaminating poly-foil pouch packaging

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.