Learn More
Waters Corp Sep-Pak Vac Cartridge, Carbon Black/Amino, 6 cc, 500 mg, 37–105 μm
Catalog No. 50781721 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This vacuum cartridge contains 500 mg of carbon black/amino propyl bonded silica, separated by a frit. It is used for pesticide cleanup from food matrices and for removing coloring from samples prior to gas chromatography (GC) analysis. This equipment is also suitable for JPMHLW official methods for propham and pesticides in food.
  • Pesticide cleanup from food matrices before GC analysis.
  • Two-layer (500 mg each) carbon black/aminopropyl bonded silica sorbent bed.
  • Removes coloring from samples.
  • Rapid sample preparation; suitable for manual or robotic use.
  • Similar behavior to reversed-phase HPLC columns.
  • Reduces costs, increases recoveries, enhances safety.
  • Improves accuracy, prevents cross-contamination and emulsion issues.
  • Easy extraction and transfer of samples.
Catalog No. 50-781-721 Supplier Waters Corp Supplier No. 186003369
This vacuum cartridge contains 500 mg of carbon black/amino propyl bonded silica, separated by a frit. It is used for pesticide cleanup from food matrices and for removing coloring from samples prior to gas chromatography (GC) analysis. This equipment is also suitable for JPMHLW official methods for propham and pesticides in food.
  • Pesticide cleanup from food matrices before GC analysis.
  • Two-layer (500 mg each) carbon black/aminopropyl bonded silica sorbent bed.
  • Removes coloring from samples.
  • Rapid sample preparation; suitable for manual or robotic use.
  • Similar behavior to reversed-phase HPLC columns.
  • Reduces costs, increases recoveries, enhances safety.
  • Improves accuracy, prevents cross-contamination and emulsion issues.
  • Easy extraction and transfer of samples.