Learn More
Waters Corp Vac Cartridge, Carbon Black/PSA Silica, 6 cc, 500 mg Sorbent
Catalog No. 502391699 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This vac cartridge features a two-layer sorbent bed designed for pesticide clean-up in food matrices prior to Gas Chromatography (GC) analysis. The PSA (Primary-Secondary Amine) sorbent provides alternative selectivity. It is a convenient, reproducible, disposable solid-phase extraction (SPE) device manufactured in an ISO 13485 and ISO 9002-registered facility, adhering to cGMP guidelines for Class 1 Medical Devices.
  • Efficient pesticide clean-up in food matrices.
  • Enhanced selectivity with PSA sorbent.
  • Compliant with JPMHLW official methods.
  • Reproducible and disposable SPE device.
  • Quality assured by ISO 13485 and ISO 9002-registered manufacturing.
  • Versatile for manual and automated SPE.
  • Optimal sorbent configuration with 37–105 μm carbon black and 37–55 μm PSA particle sizes.
  • Mass spectrometry compatible.
  • Hold-up volume of 1.4 mL.
Catalog No. 50-239-1699 Supplier Waters Corp Supplier No. 186004590
This vac cartridge features a two-layer sorbent bed designed for pesticide clean-up in food matrices prior to Gas Chromatography (GC) analysis. The PSA (Primary-Secondary Amine) sorbent provides alternative selectivity. It is a convenient, reproducible, disposable solid-phase extraction (SPE) device manufactured in an ISO 13485 and ISO 9002-registered facility, adhering to cGMP guidelines for Class 1 Medical Devices.
  • Efficient pesticide clean-up in food matrices.
  • Enhanced selectivity with PSA sorbent.
  • Compliant with JPMHLW official methods.
  • Reproducible and disposable SPE device.
  • Quality assured by ISO 13485 and ISO 9002-registered manufacturing.
  • Versatile for manual and automated SPE.
  • Optimal sorbent configuration with 37–105 μm carbon black and 37–55 μm PSA particle sizes.
  • Mass spectrometry compatible.
  • Hold-up volume of 1.4 mL.