Learn More
Waters Corp Waters Sep-Pak Accell Plus QMA Classic Cartridge, 360 mg, 37-55 μm
Catalog No. 50784988 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Accell Plus QMA Classic cartridges are silica-based, hydrophilic, strong anion-exchangers with large 300Å pores.
Catalog No. 50-784-988 Supplier Waters Corp Supplier No. WAT010835
Sep-Pak Accell Plus QMA Classic cartridges are silica-based, hydrophilic, strong anion-exchangers with large 300Å pores.