Learn More
Waters Corp Waters Sep-Pak Classic Cartridge, 360 mg Sorbent, 37 to 55 μm, 50/pk
Catalog No. 50784989 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Accell Plus CM Classic cartridges are silica-based, hydrophilic weak cation-exchangers for extracting cationic analytes. They are suitable for isolating proteins, steroids, pesticides, and herbicides.
  • Silica-based, hydrophilic weak cation-exchanger.
  • Designed for extraction of cationic analytes.
  • Ideal for isolation of large molecules, like cationic proteins.
  • Can extract steroids, pesticides, and herbicides.
  • Use with Luer-tip syringes or vacuum-assisted devices.
  • Shipped sealed in a non-contaminating poly-foil pouch.
  • Ion exchange capacity: 370 μeq/gram.
  • Mass spec compatible.
  • Water wettable.
Catalog No. 50-784-989 Supplier Waters Corp Supplier No. WAT010910
Sep-Pak Accell Plus CM Classic cartridges are silica-based, hydrophilic weak cation-exchangers for extracting cationic analytes. They are suitable for isolating proteins, steroids, pesticides, and herbicides.
  • Silica-based, hydrophilic weak cation-exchanger.
  • Designed for extraction of cationic analytes.
  • Ideal for isolation of large molecules, like cationic proteins.
  • Can extract steroids, pesticides, and herbicides.
  • Use with Luer-tip syringes or vacuum-assisted devices.
  • Shipped sealed in a non-contaminating poly-foil pouch.
  • Ion exchange capacity: 370 μeq/gram.
  • Mass spec compatible.
  • Water wettable.