Learn More
Waters Corp Waters Corporation Solid Phase Extraction Cartridge, Alumina A, 1850 mg, 50–300 μm
Catalog No. 50785653 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Alumina A Classic cartridges feature a highly active grade of alumina with an acidic surface chemistry, providing an extremely polar surface for analyte retention and acting as a Lewis acid. This normal phase sorbent is comparable to silica in use but offers greater stability under high pH conditions. It also functions as a low-capacity ion exchanger in aqueous media and is unaffected by high-energy radioactivity, making it suitable for isolating radioactive compounds. These Classic-style cartridges are designed for positive displacement with a Luer-tip syringe and can be easily attached to vacuum-assisted flow devices using a male/male adaptor.
  • Highly active alumina with acidic surface chemistry.
  • More stable than silica under high pH conditions.
  • Unaffected by high-energy radioactivity.
  • Designed for positive displacement by Luer-tip syringe.
Catalog No. 50-785-653 Supplier Waters Corp Supplier No. WAT051800
Sep-Pak Alumina A Classic cartridges feature a highly active grade of alumina with an acidic surface chemistry, providing an extremely polar surface for analyte retention and acting as a Lewis acid. This normal phase sorbent is comparable to silica in use but offers greater stability under high pH conditions. It also functions as a low-capacity ion exchanger in aqueous media and is unaffected by high-energy radioactivity, making it suitable for isolating radioactive compounds. These Classic-style cartridges are designed for positive displacement with a Luer-tip syringe and can be easily attached to vacuum-assisted flow devices using a male/male adaptor.
  • Highly active alumina with acidic surface chemistry.
  • More stable than silica under high pH conditions.
  • Unaffected by high-energy radioactivity.
  • Designed for positive displacement by Luer-tip syringe.