Learn More
Waters Corp Sep-Pak Classic Cartridge, Alumina B, 1850 mg Sorbent, 50 – 300 μm
Catalog No. 50785655 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Alumina B Classic cartridges utilize highly active alumina with a basic surface chemistry for analyte retention and as a low-capacity ion exchanger.
  • Highly active alumina with basic surface chemistry for polar retention and Lewis acid properties.
  • Normal phase sorbent, stable under high pH; also acts as a low-capacity ion exchanger.
  • Extracts phospholipids, steroids, and catecholamines; unaffected by high-energy radioactivity.
  • Classic-style cartridges designed for Luer-tip syringe use and vacuum-assisted flow via adaptor.
Catalog No. 50-785-655 Supplier Waters Corp Supplier No. WAT051820
Sep-Pak Alumina B Classic cartridges utilize highly active alumina with a basic surface chemistry for analyte retention and as a low-capacity ion exchanger.
  • Highly active alumina with basic surface chemistry for polar retention and Lewis acid properties.
  • Normal phase sorbent, stable under high pH; also acts as a low-capacity ion exchanger.
  • Extracts phospholipids, steroids, and catecholamines; unaffected by high-energy radioactivity.
  • Classic-style cartridges designed for Luer-tip syringe use and vacuum-assisted flow via adaptor.