Learn More
Waters Corp Waters Sep-Pak Classic Cartridge, C18, 360 mg, 55–105 μm
Catalog No. 50785971 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This silica-based bonded phase with strong hydrophobicity is designed to adsorb analytes, even those with weak hydrophobicity, from aqueous solutions. It is similar in behavior to reversed-phase HPLC columns.
  • Silica-based bonded phase with strong hydrophobicity.
  • Used to adsorb analytes of even weak hydrophobicity from aqueous solutions.
  • Typical applications include drugs and their metabolites in serum, plasma or urine, desalting of peptides, trace organics in environmental water samples, and organic acids in beverages.
  • Similar in behavior to reversed-phase HPLC columns.
Catalog No. 50-785-971 Supplier Waters Corp Supplier No. WAT091139
Add to Cart
Add to Cart
Request a Quote
This silica-based bonded phase with strong hydrophobicity is designed to adsorb analytes, even those with weak hydrophobicity, from aqueous solutions. It is similar in behavior to reversed-phase HPLC columns.
  • Silica-based bonded phase with strong hydrophobicity.
  • Used to adsorb analytes of even weak hydrophobicity from aqueous solutions.
  • Typical applications include drugs and their metabolites in serum, plasma or urine, desalting of peptides, trace organics in environmental water samples, and organic acids in beverages.
  • Similar in behavior to reversed-phase HPLC columns.