Learn More
Waters Corp Sep-Pak Cyanopropyl (CN) Classic Cartridge, 360 mg Sorbent per Cartridge, 55 - 105 µm, 50/pk
Supplier:  Waters Corp WAT010823
Encompass_Preferred
Silica-based polar bonded phase; can be used as less polar alternative to silica in normal-phase applications or as less hydrophobic alternative to C18 or C8 in reversed-phase applications; typical applications include drug metabolites and pesticides.
Catalog No. 50-784-986