Learn More
Waters Corp Sep-Pak Aminopropyl (NH2) Classic Cartridge, 360 mg, 55 to 105 μm
Catalog No. 50784987 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Sep-Pak Aminopropyl (NH2) Classic cartridges provide a moderately polar, silica-based bonded phase with a weakly basic surface. They function as polar sorbents or weak anion exchangers (below pH 8), ideal for extracting various analytes including phenols, saccharides, drugs, and metabolites.
  • Moderately polar, silica-based bonded phase with a weakly basic surface
  • Functions as a polar sorbent or weak anion exchanger (below pH 8)
  • Suitable for extracting phenols, phenolic pigments, petroleum fractionation, saccharides, drugs, and metabolites
  • Designed for positive displacement with Luer-tip syringes
  • Attachable to vacuum-assisted flow devices with a male/male adaptor
Catalog No. 50-784-987 Supplier Waters Corp Supplier No. WAT010830
These Sep-Pak Aminopropyl (NH2) Classic cartridges provide a moderately polar, silica-based bonded phase with a weakly basic surface. They function as polar sorbents or weak anion exchangers (below pH 8), ideal for extracting various analytes including phenols, saccharides, drugs, and metabolites.
  • Moderately polar, silica-based bonded phase with a weakly basic surface
  • Functions as a polar sorbent or weak anion exchanger (below pH 8)
  • Suitable for extracting phenols, phenolic pigments, petroleum fractionation, saccharides, drugs, and metabolites
  • Designed for positive displacement with Luer-tip syringes
  • Attachable to vacuum-assisted flow devices with a male/male adaptor