Learn More
Waters Corp Sep-Pak Silica Classic Cartridge, 690 mg Sorbent per Cartridge, 55 - 105 µm, 2000/pk
Catalog No. 50785972 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Polar sorbent, used primarily to adsorb analytes from non polar solvents like hydrocarbons, chloro- or flouro-substituted hydrocarbons or less polar esters and ethers.
Catalog No. 50-785-972 Supplier Waters Corp Supplier No. WAT091197
Add to Cart
Add to Cart
Request a Quote
Polar sorbent, used primarily to adsorb analytes from non polar solvents like hydrocarbons, chloro- or flouro-substituted hydrocarbons or less polar esters and ethers.