Learn More
Waters Corp Waters Sep-Pak DNPH-Silica Cartridge, 350 mg, 55 to 105 μm
Catalog No. 50785423 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These cartridges are designed for analyzing formaldehyde, aldehydes, and ketones in air, meeting EPA Method TO-11A, JPM HLW, and ISO 16000 requirements. They trap these compounds by reacting them with DNPH to form stable hydrazone derivatives during sample collection.
  • Analyzes formaldehyde, aldehydes, and ketones in air, meeting EPA TO-11A, JPM HLW, and ISO 16000.
  • Utilizes vacuum pump for air sample collection and hydrazone derivative retention for HPLC/UPLC quantitation.
  • Offers high-speed, high-resolution analysis for low parts-per-billion quantitation.
  • Minimizes laboratory costs through reduced solvent consumption and hazardous waste disposal.
  • Ideal for field sampling and process monitoring with >95% collection efficiency.
Catalog No. 50-785-423 Supplier Waters Corp Supplier No. WAT037500
These cartridges are designed for analyzing formaldehyde, aldehydes, and ketones in air, meeting EPA Method TO-11A, JPM HLW, and ISO 16000 requirements. They trap these compounds by reacting them with DNPH to form stable hydrazone derivatives during sample collection.
  • Analyzes formaldehyde, aldehydes, and ketones in air, meeting EPA TO-11A, JPM HLW, and ISO 16000.
  • Utilizes vacuum pump for air sample collection and hydrazone derivative retention for HPLC/UPLC quantitation.
  • Offers high-speed, high-resolution analysis for low parts-per-billion quantitation.
  • Minimizes laboratory costs through reduced solvent consumption and hazardous waste disposal.
  • Ideal for field sampling and process monitoring with >95% collection efficiency.