Learn More
Waters Corp Waters Sep-Pak Cartridge, DNPH-Silica, 800 mg, 55–105 μm, 20/pk
Catalog No. 50785486 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sep-Pak DNPH-Silica Plus Long Cartridge is designed for analyzing formaldehyde, other aldehydes, and ketones in air. It meets EPA Method TO-11A, JPM HLW, and ISO 16000 requirements, making it an excellent device for sample collection. Air is redirected through the cartridge using a vacuum pump, allowing ketones and aldehydes to react with DNPH, forming hydrazone derivatives. These hydrazones can then be eluted with acetonitrile for sample analysis.
  • Provides low background for higher sensitivity.
  • Saves time and increases productivity.
  • Offers superior convenience and reproducibility, ideal for field sampling and process monitoring.
  • Reduces solvent consumption, solvent exposure, and hazardous waste disposal costs.
  • Supports high-speed, high-resolution UPLC Technology applications for excellent quantitation capability in the low parts-per-billion range.
Catalog No. 50-785-486 Supplier Waters Corp Supplier No. WAT039550
The Sep-Pak DNPH-Silica Plus Long Cartridge is designed for analyzing formaldehyde, other aldehydes, and ketones in air. It meets EPA Method TO-11A, JPM HLW, and ISO 16000 requirements, making it an excellent device for sample collection. Air is redirected through the cartridge using a vacuum pump, allowing ketones and aldehydes to react with DNPH, forming hydrazone derivatives. These hydrazones can then be eluted with acetonitrile for sample analysis.
  • Provides low background for higher sensitivity.
  • Saves time and increases productivity.
  • Offers superior convenience and reproducibility, ideal for field sampling and process monitoring.
  • Reduces solvent consumption, solvent exposure, and hazardous waste disposal costs.
  • Supports high-speed, high-resolution UPLC Technology applications for excellent quantitation capability in the low parts-per-billion range.