Learn More
Waters Corp Sep-Pak Solid Phase Extraction Cartridge, tC18, 900 mg, 37 to 55 μm
Catalog No. 50785397 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This silica-based bonded phase product features strong hydrophobicity and trifunctional bonding chemistry for increased hydrolytic stability. It simplifies protocols by reducing steps and minimizing sample handling, suitable for robust performance in diverse applications.
  • Silica-based bonded phase with strong hydrophobicity.
  • Trifunctional bonding chemistry provides increased hydrolytic stability.
  • Simplifies protocols and reduces sample handling.
  • Helps avoid emulsion issues.
  • Improves safety by reducing harmful agent exposure.
  • Enables direct field sampling.
  • Features a high-purity polypropylene syringe-barrel body with polyethylene frits.
  • Utilizes with multi-position vacuum manifolds via a standard male-Luer tip.
  • Suitable for analyte isolation and enrichment from biological, environmental, and dietary samples.
Catalog No. 50-785-397 Supplier Waters Corp Supplier No. WAT036800
This silica-based bonded phase product features strong hydrophobicity and trifunctional bonding chemistry for increased hydrolytic stability. It simplifies protocols by reducing steps and minimizing sample handling, suitable for robust performance in diverse applications.
  • Silica-based bonded phase with strong hydrophobicity.
  • Trifunctional bonding chemistry provides increased hydrolytic stability.
  • Simplifies protocols and reduces sample handling.
  • Helps avoid emulsion issues.
  • Improves safety by reducing harmful agent exposure.
  • Enables direct field sampling.
  • Features a high-purity polypropylene syringe-barrel body with polyethylene frits.
  • Utilizes with multi-position vacuum manifolds via a standard male-Luer tip.
  • Suitable for analyte isolation and enrichment from biological, environmental, and dietary samples.