Learn More
Waters Corp Sep-Pak Accell Plus QMA Plus Light Cartridge, 130 mg Sorbent per Cartridge, 37 - 55 µm, 50/pk
Catalog No. 50785179 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Accell Plus QMA Plus Light cartridges contain a silica-based, hydrophilic, strong anion-exchanger with large pore size (300Å). The cartridges are designed for extraction of anionic analytes in aqueous and non-aqueous solutions. The large pore size makes it ideal for isolation of larger molecules such as anionic proteins such as immunoglobulins and enzymes. Other applications include isolation of phenolic compounds and removal of acidic pigments in wine, fruit juices, and food extracts. These cartridges have a distinctive finned outer body and a reduced internal diameter, which results in an interstitial volume about one-third that of the corresponding Plus-style cartridge. This design allows you to elute fractions in a minimal volume to improve recoveries and reduce solvent consumption, a benefit especially for applications where samples are limited or where excessive dilution is a concern.
Catalog No. 50-785-179 Supplier Waters Corp Supplier No. WAT023525
Sep-Pak Accell Plus QMA Plus Light cartridges contain a silica-based, hydrophilic, strong anion-exchanger with large pore size (300Å). The cartridges are designed for extraction of anionic analytes in aqueous and non-aqueous solutions. The large pore size makes it ideal for isolation of larger molecules such as anionic proteins such as immunoglobulins and enzymes. Other applications include isolation of phenolic compounds and removal of acidic pigments in wine, fruit juices, and food extracts. These cartridges have a distinctive finned outer body and a reduced internal diameter, which results in an interstitial volume about one-third that of the corresponding Plus-style cartridge. This design allows you to elute fractions in a minimal volume to improve recoveries and reduce solvent consumption, a benefit especially for applications where samples are limited or where excessive dilution is a concern.