Learn More
Waters Corp Waters Corporation Sep-Pak Accell Plus QMA Carbonate Plus Light Cartridge, 46 mg Sorbent, 40 μm, 50 pk
Catalog No. 50466013 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These cartridges feature a silica-based, hydrophilic, strong anion-exchanger with a large pore size (300Å) and a carbonate counter-ion. They are designed for the extraction of anionic analytes from various solutions, especially where chloride interference is a concern.
  • Silica-based, hydrophilic, strong anion-exchanger with 300Å pore size and carbonate counter-ion.
  • Extracts anionic analytes from aqueous/non-aqueous solutions; avoids chloride interference.
  • Light-style format with reduced internal diameter; enables minimal volume elution.
  • Improves recoveries and reduces solvent consumption for limited samples.
  • Ideal for isolating anionic proteins, acidic pigments, phenolic compounds, and inorganic anions.
  • Manufactured in ISO 13485/9002-registered facility, compliant with cGMP for Class 1 Medical Devices.
Catalog No. 50-466-013 Supplier Waters Corp Supplier No. 186004540
These cartridges feature a silica-based, hydrophilic, strong anion-exchanger with a large pore size (300Å) and a carbonate counter-ion. They are designed for the extraction of anionic analytes from various solutions, especially where chloride interference is a concern.
  • Silica-based, hydrophilic, strong anion-exchanger with 300Å pore size and carbonate counter-ion.
  • Extracts anionic analytes from aqueous/non-aqueous solutions; avoids chloride interference.
  • Light-style format with reduced internal diameter; enables minimal volume elution.
  • Improves recoveries and reduces solvent consumption for limited samples.
  • Ideal for isolating anionic proteins, acidic pigments, phenolic compounds, and inorganic anions.
  • Manufactured in ISO 13485/9002-registered facility, compliant with cGMP for Class 1 Medical Devices.