Learn More
Waters Corp Sep-Pak Accell Plus QMA Carbonate Plus Light Cartridge, 130 mg Sorbent per Cartridge, 37 - 55 µm, 50/pk
Catalog No. 50465999 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Accell Plus QMA Carbonate Plus Light cartridges contain a silica-based, hydrophilic, strong anion-exchanger with a large pore size (300Å) and a carbonate counter-ion. These cartridges are used for extraction of anionic analytes in aqueous and non-aqueous solutions when the chloride counter-ion in the standard Sep-Pak Accell Plus QMA cartridges interferes with the analysis. These Light-style cartridges have a distinctive finned outer body and a reduced internal diameter, which results in an interstitial volume about one-third that of the corresponding Plus-style cartridge. This design allows you to elute fractions in a minimal volume to improve recoveries and reduce solvent consumption, a benefit especially for applications where samples are limited or where excessive dilution is a concern.
Catalog No. 50-465-999 Supplier Waters Corp Supplier No. 186004051
Sep-Pak Accell Plus QMA Carbonate Plus Light cartridges contain a silica-based, hydrophilic, strong anion-exchanger with a large pore size (300Å) and a carbonate counter-ion. These cartridges are used for extraction of anionic analytes in aqueous and non-aqueous solutions when the chloride counter-ion in the standard Sep-Pak Accell Plus QMA cartridges interferes with the analysis. These Light-style cartridges have a distinctive finned outer body and a reduced internal diameter, which results in an interstitial volume about one-third that of the corresponding Plus-style cartridge. This design allows you to elute fractions in a minimal volume to improve recoveries and reduce solvent consumption, a benefit especially for applications where samples are limited or where excessive dilution is a concern.