Learn More
Waters Corp Waters Sep-Pak Cartridge, Alumina A, 280 mg Sorbent, 50-300 μm, Plus Light
Catalog No. 50785183 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These cartridges feature a highly active grade of alumina with an acidic surface chemistry, providing an extremely polar surface for analyte retention. They are a normal phase sorbent, suitable for radioactive compound isolation, and designed for efficient elution with minimal solvent.
  • Highly active alumina with acidic surface chemistry.
  • Extremely polar surface for analyte retention.
  • Normal phase sorbent, comparable to silica, stable under high pH.
  • Low-capacity ion exchanger in aqueous media.
  • Resistant to high-energy radioactivity.
  • Reduced internal diameter and distinctive finned outer body.
  • Minimal elution volume, improving recoveries and reducing solvent consumption.
Catalog No. 50-785-183 Supplier Waters Corp Supplier No. WAT023549
These cartridges feature a highly active grade of alumina with an acidic surface chemistry, providing an extremely polar surface for analyte retention. They are a normal phase sorbent, suitable for radioactive compound isolation, and designed for efficient elution with minimal solvent.
  • Highly active alumina with acidic surface chemistry.
  • Extremely polar surface for analyte retention.
  • Normal phase sorbent, comparable to silica, stable under high pH.
  • Low-capacity ion exchanger in aqueous media.
  • Resistant to high-energy radioactivity.
  • Reduced internal diameter and distinctive finned outer body.
  • Minimal elution volume, improving recoveries and reducing solvent consumption.