Learn More
Waters Corp Sep-Pak Cartridge, Alumina N, 280 mg, 50-300 μm
Catalog No. 50785185 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Alumina N Plus Light cartridges feature a highly active alumina sorbent with neutral surface chemistry for normal-phase solid phase extraction. This sorbent provides an extremely polar surface for analyte retention, similar to silica but more stable under high pH conditions. Its unique design minimizes interstitial volume, allowing efficient elution in minimal solvent and optimizing recoveries.
  • Highly active alumina sorbent with neutral chemistry.
  • Extremely polar surface for effective analyte retention.
  • More stable than unfunctionalized silica under high pH.
  • Suitable for synthetic crude oil fractionation and similar uses.
  • Reduced interstitial volume vs. Plus-style cartridges.
  • Enables elution of fractions in minimal volume, improving recoveries.
  • Reduces solvent consumption, beneficial for limited samples.
Catalog No. 50-785-185 Supplier Waters Corp Supplier No. WAT023561
Sep-Pak Alumina N Plus Light cartridges feature a highly active alumina sorbent with neutral surface chemistry for normal-phase solid phase extraction. This sorbent provides an extremely polar surface for analyte retention, similar to silica but more stable under high pH conditions. Its unique design minimizes interstitial volume, allowing efficient elution in minimal solvent and optimizing recoveries.
  • Highly active alumina sorbent with neutral chemistry.
  • Extremely polar surface for effective analyte retention.
  • More stable than unfunctionalized silica under high pH.
  • Suitable for synthetic crude oil fractionation and similar uses.
  • Reduced interstitial volume vs. Plus-style cartridges.
  • Enables elution of fractions in minimal volume, improving recoveries.
  • Reduces solvent consumption, beneficial for limited samples.