Learn More
Waters Corp Waters Sep-Pak, C18 Plus Light, 130 mg Sorbent, 55 to 105 um
Catalog No. 50785175 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Sep-Pak C18 Plus Light Cartridge is a silica-based bonded phase designed for efficient sample enrichment and purification. It effectively adsorbs weakly hydrophobic analytes from aqueous solutions, behaving similarly to reversed-phase HPLC columns.
  • Silica-based bonded phase with strong hydrophobicity.
  • Adsorbs weakly hydrophobic analytes.
  • Ideal for sample enrichment and purification.
  • High-quality packing materials for optimal surface area and pore-size.
  • Features a 20 μm frit for solvent resistance, low extractables, good flow properties, and depth filtration.
  • Sealed in a poly-foil pouch to maintain product integrity and purity.
  • Optimized for a pH range of 2 to 8.
  • Contains 130 mg of sorbent for lighter sample loads.
Catalog No. 50-785-175 Supplier Waters Corp Supplier No. WAT023501
This Sep-Pak C18 Plus Light Cartridge is a silica-based bonded phase designed for efficient sample enrichment and purification. It effectively adsorbs weakly hydrophobic analytes from aqueous solutions, behaving similarly to reversed-phase HPLC columns.
  • Silica-based bonded phase with strong hydrophobicity.
  • Adsorbs weakly hydrophobic analytes.
  • Ideal for sample enrichment and purification.
  • High-quality packing materials for optimal surface area and pore-size.
  • Features a 20 μm frit for solvent resistance, low extractables, good flow properties, and depth filtration.
  • Sealed in a poly-foil pouch to maintain product integrity and purity.
  • Optimized for a pH range of 2 to 8.
  • Contains 130 mg of sorbent for lighter sample loads.