Learn More
Waters Corp Waters Sep-Pak C8 Plus Light Cartridge, 145 mg, 50 Pack
Catalog No. 50785391 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sep-Pak C8 Plus Light Cartridge is a precision chromatographic instrument designed for sample enrichment and purification. It features a silica-based bonded octadecyl phase with moderate hydrophobicity, ideal for adsorbing analytes from aqueous solutions.
  • Utilizes a silica-based, endcapped, reversed-phase C8 bonded phase with moderate hydrophobicity.
  • Contains 145 mg of sorbent material with a particle size of 37 – 55 μm.
  • Packaged as 50 units per box.
  • Features a pore size of 125 Å on a silica sorbent substrate.
  • Offers a pH range of 2 to 8 and a hold up volume of 0.4 mL.
  • Sealed in a non-contaminating poly-foil bag to ensure product integrity and purity.
  • Equipped with a 20 μm frit for good flow, solvent resistance, low extractables, and depth filtration.
Catalog No. 50-785-391 Supplier Waters Corp Supplier No. WAT036770
The Sep-Pak C8 Plus Light Cartridge is a precision chromatographic instrument designed for sample enrichment and purification. It features a silica-based bonded octadecyl phase with moderate hydrophobicity, ideal for adsorbing analytes from aqueous solutions.
  • Utilizes a silica-based, endcapped, reversed-phase C8 bonded phase with moderate hydrophobicity.
  • Contains 145 mg of sorbent material with a particle size of 37 – 55 μm.
  • Packaged as 50 units per box.
  • Features a pore size of 125 Å on a silica sorbent substrate.
  • Offers a pH range of 2 to 8 and a hold up volume of 0.4 mL.
  • Sealed in a non-contaminating poly-foil bag to ensure product integrity and purity.
  • Equipped with a 20 μm frit for good flow, solvent resistance, low extractables, and depth filtration.