Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Sep-Pak Cartridge, Diol, 130 mg, 37 to 55 μm

Catalog No. 50785178 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This silica-based polar bonded phase cartridge has a neutral character. It can be used as an alternative to silica in normal-phase applications where the acidic character of silica is undesirable, or as a very weakly interacting phase in aqueous applications. Key applications include the isolation of antibiotics from cosmetics and the isolation of proteins or peptides by hydrophobic interaction chromatography.

  • Diol chemistry
  • Not endcapped
  • Mass spectrometer compatible
  • Reversed phase mode
  • Particle size of 37 to 55 μm
  • Pore size of 300 Å
  • Silica sorbent substrate
  • Sorbent weight of 130 mg
  • Not water wettable
  • pH range of 2 to 8
  • Hold up volume of 0.3 mL

Catalog No. 50-785-178 Supplier Waters Corp Supplier No. WAT023519
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This silica-based polar bonded phase cartridge has a neutral character. It can be used as an alternative to silica in normal-phase applications where the acidic character of silica is undesirable, or as a very weakly interacting phase in aqueous applications. Key applications include the isolation of antibiotics from cosmetics and the isolation of proteins or peptides by hydrophobic interaction chromatography.

  • Diol chemistry
  • Not endcapped
  • Mass spectrometer compatible
  • Reversed phase mode
  • Particle size of 37 to 55 μm
  • Pore size of 300 Å
  • Silica sorbent substrate
  • Sorbent weight of 130 mg
  • Not water wettable
  • pH range of 2 to 8
  • Hold up volume of 0.3 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.