Learn More
Waters Corp Waters Solid Phase Extraction Cartridge, Florisil, 145 mg, 50-200 μm
Catalog No. 50785182 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This solid phase extraction cartridge features a polar, highly active, weakly basic sorbent. It is designed for the adsorption of low to moderate polarity species from nonaqueous solutions.
  • Polar, highly active, weakly basic sorbent
  • Designed for adsorption of low to moderate polarity species from nonaqueous solutions
  • Mass spectrometry compatible
Catalog No. 50-785-182 Supplier Waters Corp Supplier No. WAT023543
This solid phase extraction cartridge features a polar, highly active, weakly basic sorbent. It is designed for the adsorption of low to moderate polarity species from nonaqueous solutions.
  • Polar, highly active, weakly basic sorbent
  • Designed for adsorption of low to moderate polarity species from nonaqueous solutions
  • Mass spectrometry compatible