Learn More
Waters Corp Sep-Pak Aminopropyl (NH2) Plus Light Cartridge, 130 mg Sorbent, 55–105 μm
Catalog No. 50785177 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Amino Propyl (NH2) Plus Light cartridges contain a moderately polar, silica-based bonded phase with a weakly basic surface. The sorbent can be used as a polar sorbent with different selectivity for acidic/basic analytes or as a weak anion exchanger in aqueous medium below pH 8. Applications include the extraction of phenols and phenolic pigments, petroleum fractionation, saccharides, and drugs and metabolites.
  • Finned outer body and reduced internal diameter for improved interstitial volume.
  • Allows elution of fractions in minimal volume to improve recoveries and reduce solvent consumption.
  • Sealed within a non-contaminating poly-foil pouch to maintain product integrity.
  • Optimal particle size distribution, pore size, and surface area for uniform results.
Catalog No. 50-785-177 Supplier Waters Corp Supplier No. WAT023513
Sep-Pak Amino Propyl (NH2) Plus Light cartridges contain a moderately polar, silica-based bonded phase with a weakly basic surface. The sorbent can be used as a polar sorbent with different selectivity for acidic/basic analytes or as a weak anion exchanger in aqueous medium below pH 8. Applications include the extraction of phenols and phenolic pigments, petroleum fractionation, saccharides, and drugs and metabolites.
  • Finned outer body and reduced internal diameter for improved interstitial volume.
  • Allows elution of fractions in minimal volume to improve recoveries and reduce solvent consumption.
  • Sealed within a non-contaminating poly-foil pouch to maintain product integrity.
  • Optimal particle size distribution, pore size, and surface area for uniform results.