Learn More
Waters Corp Sep-Pak Silica Plus Light Cartridge, 120 mg Sorbent per Cartridge, 55 - 105 µm, 50/pk
Catalog No. 50785181 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Polar sorbent, used primarily to adsorb analytes from non polar solvents like hydrocarbons, chloro- or flouro-substituted hydrocarbons or less polar esters and ethers.
Catalog No. 50-785-181 Supplier Waters Corp Supplier No. WAT023537
Polar sorbent, used primarily to adsorb analytes from non polar solvents like hydrocarbons, chloro- or flouro-substituted hydrocarbons or less polar esters and ethers.